Learn More
Pyramex Safety Black Reflective Safety Vest
Catalog No. 19156148 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-156-148 5X-Large
19-156-136 Small
19-156-140 Medium
19-156-143 Large
19-156-144 X-Large
19-156-145 2X-Large
19-156-146 3X-Large
19-156-147 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-156-148 Supplier Pyramex Safety Supplier No. RVZ2411CPX5
Add to Cart
Add to Cart
Pyramex Black Reflective Safety Vest features include high visibility markings on the front and back, pockets, and a zipper front closure.
  • Lightweight polyester solid material on front, mesh material on back
  • 2 in. silver reflective material with 4 in. contrasting color trim
  • Clear front pocket
  • 8 pockets: 5 outside, 3 inside
  • Badge/Microphone tabs located 3 in. below each shoulder seam
Specifications
Closure Type Zipper
Certifications/Compliance ANSI/ISEA 107-2015 Type 0 Class 1 Not FR
Color Black
Size 5X-Large
Material Polyester
Tape Type Generic Silver Reflective