Learn More
AVANTOR MASTERFLEX (VWR LLC) Black Rubber Vacuum Tubing, 3/8"" ID x 7/8"" OD, 25ft. 96143-08
Catalog No. NC1200666
Encompass
Applications: Vacuum and pressure applications for general lab and industrial use Characteristics: Thick wall, flexible, opaque Temperature Range: 0¿F to 150¿F Sterilize: Ethylene oxide or autoclave for 30 minutes at 15 psi
Catalog No. NC1200666 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9614308
Applications: Vacuum and pressure applications for general lab and industrial use Characteristics: Thick wall, flexible, opaque Temperature Range: 0¿F to 150¿F Sterilize: Ethylene oxide or autoclave for 30 minutes at 15 psi