Learn More
MSC CRC 1004799 8 oz Tube Black RTV Silicone Gasket Sealant -30 to 450°F Operating Temp, 24 hr Full Cure Time
Catalog No. 501304740
Encompass
CRC 1004799 8 oz Tube Black RTV Silicone Gasket Sealant -30 to 450°F Operating Temp, 24 hr Full Cure Time
Catalog No. 50-130-4740 Supplier MSC Supplier No. 01292275
CRC 1004799 8 oz Tube Black RTV Silicone Gasket Sealant -30 to 450°F Operating Temp, 24 hr Full Cure Time