Learn More
MSC Black Stallion FR Welding Jacket, Royal Blue, Large, 1 ea
Catalog No. 5000803322
Encompass
This contoured FR cotton welding jacket offers a modern design for a better fitting, better looking, and more comfortable protective garment, providing a new level of innovation for welders.
  • Made with 9 oz. FR-treated cotton for lightweight protection from heat, sparks, and spatter
  • Welder's collar with adjustable snap options protects the neck against sparks
  • Adjustable waist straps with two snap positions for a customized fit and trimmer appearance
  • Extended length front coverage offers added protection
  • Dual scribe pockets – one on each sleeve
  • Convenient snap front and wrists
  • Leather reinforcements on all snaps
  • Zippered inside pocket
Catalog No. 50-008-03322 Supplier MSC Supplier No. 53678660
This contoured FR cotton welding jacket offers a modern design for a better fitting, better looking, and more comfortable protective garment, providing a new level of innovation for welders.
  • Made with 9 oz. FR-treated cotton for lightweight protection from heat, sparks, and spatter
  • Welder's collar with adjustable snap options protects the neck against sparks
  • Adjustable waist straps with two snap positions for a customized fit and trimmer appearance
  • Extended length front coverage offers added protection
  • Dual scribe pockets – one on each sleeve
  • Convenient snap front and wrists
  • Leather reinforcements on all snaps
  • Zippered inside pocket