Learn More
MSC Black Stallion FR Welding Jacket, Royal Blue, S-5XL, 1 ea
Catalog No. 5000684965
Encompass
The BSX Contoured FR Cotton Welding Jacket in Royal Blue offers improved fit, appearance, and comfort while providing essential protection. Designed for welders, this jacket shields against heat, sparks, and spatter.
  • Provides lightweight protection with 9 oz. FR-treated cotton.
  • Welder's collar with adjustable snap options protects neck against sparks.
  • Adjustable waist straps with two snap positions for a customized fit.
  • Extended length front coverage offers added protection.
  • Dual scribe pockets, one on each sleeve, and a zippered inside pocket.
  • Convenient snap front and wrists.
  • Leather reinforcements on all snaps.
  • Available in sizes S-5XL.
Catalog No. 50-006-84965 Supplier MSC Supplier No. 73426546
The BSX Contoured FR Cotton Welding Jacket in Royal Blue offers improved fit, appearance, and comfort while providing essential protection. Designed for welders, this jacket shields against heat, sparks, and spatter.
  • Provides lightweight protection with 9 oz. FR-treated cotton.
  • Welder's collar with adjustable snap options protects neck against sparks.
  • Adjustable waist straps with two snap positions for a customized fit.
  • Extended length front coverage offers added protection.
  • Dual scribe pockets, one on each sleeve, and a zippered inside pocket.
  • Convenient snap front and wrists.
  • Leather reinforcements on all snaps.
  • Available in sizes S-5XL.