Learn More
MSC Black Stallion FR Wraparound Helmet Bib, 1 Dozen
Catalog No. 5000688606
Encompass
The FR Wraparound Helmet Bib is a lightweight, double-layer flame-resistant cotton helmet bib that provides wrap-around coverage to block sparks, heat, and UV rays. It features an extended front for extra protection and to reduce chest glare.
  • Double layer 7 oz. flame-resistant cotton protects against light sparks and UV
  • Reversible design offers FR protection on both sides
  • Wrap around coverage blocks more sparks, heat, and UV rays
  • Extended front coverage provides extra protection and reduces chest glare
  • Six push-on clips provide secure hold on most welding helmets
  • Quickly and easily attaches to most welding helmets
  • Sold by the dozen in innovative packaging that can be used as a counter-top display for fast sales
Catalog No. 50-006-88606 Supplier MSC Supplier No. 94594611
The FR Wraparound Helmet Bib is a lightweight, double-layer flame-resistant cotton helmet bib that provides wrap-around coverage to block sparks, heat, and UV rays. It features an extended front for extra protection and to reduce chest glare.
  • Double layer 7 oz. flame-resistant cotton protects against light sparks and UV
  • Reversible design offers FR protection on both sides
  • Wrap around coverage blocks more sparks, heat, and UV rays
  • Extended front coverage provides extra protection and reduces chest glare
  • Six push-on clips provide secure hold on most welding helmets
  • Quickly and easily attaches to most welding helmets
  • Sold by the dozen in innovative packaging that can be used as a counter-top display for fast sales