Learn More
MSC Black Stallion Leather Cape Sleeve, Split Cowhide, Large, 1 EA
Catalog No. 5000769497
Encompass
The Black Stallion Leather Cape Sleeve offers durable, heat-resistant protection for professional welders. Crafted from premium leather, it shields shoulders and upper arms from sparks, spatter, and abrasion without restricting motion. Reinforced seams and an adjustable fit ensure lasting durability and comfort over workwear, making it ideal for industrial welding tasks.
  • Durable split cowhide construction
  • Satin-lined shoulders; gusseted underarms
  • Dual scribe pockets; Kevlar® stitching
  • Covers shoulders and arms with full mobility
  • Reinforced seams for prolonged industrial wear
  • Adjustable fit for comfort and layering
Catalog No. 50-007-69497 Supplier MSC Supplier No. 58773425
The Black Stallion Leather Cape Sleeve offers durable, heat-resistant protection for professional welders. Crafted from premium leather, it shields shoulders and upper arms from sparks, spatter, and abrasion without restricting motion. Reinforced seams and an adjustable fit ensure lasting durability and comfort over workwear, making it ideal for industrial welding tasks.
  • Durable split cowhide construction
  • Satin-lined shoulders; gusseted underarms
  • Dual scribe pockets; Kevlar® stitching
  • Covers shoulders and arms with full mobility
  • Reinforced seams for prolonged industrial wear
  • Adjustable fit for comfort and layering