Learn More
MSC Black Stallion MIG Glove, Pigskin and Cowhide, 1 ea
Catalog No. 5000673904
Encompass
This glove offers a value-conscious combination of dexterity and durability, featuring a premium grain pigskin palm and a cowhide back.
  • Fully lined and padded pigskin palm for comfort with high tactile performance.
  • Exclusive DragPatch® offers extra hot zone protection.
  • Exclusive RestPatch® provides enhanced wrist padding.
  • Kevlar® stitching provides spark and abrasion resistance.
  • Available in sizes S-3XL.
Catalog No. 50-006-73904 Supplier MSC Supplier No. 55213243
This glove offers a value-conscious combination of dexterity and durability, featuring a premium grain pigskin palm and a cowhide back.
  • Fully lined and padded pigskin palm for comfort with high tactile performance.
  • Exclusive DragPatch® offers extra hot zone protection.
  • Exclusive RestPatch® provides enhanced wrist padding.
  • Kevlar® stitching provides spark and abrasion resistance.
  • Available in sizes S-3XL.