Learn More
MSC Black Stallion TruGuard 200 FR Cotton Welding Jacket, 1 ea
Catalog No. 5000675982
Encompass
This welding jacket offers reliable protection for light welding applications.
  • Constructed from 9 oz. flame-resistant treated cotton
  • Welder's collar for enhanced protection against sparks and splatter
  • Equipped with 6 convenient snap buttons, including a collar button
  • Leather reinforcements applied to all snaps
  • Scribe pocket holds welding pencils, soapstone, and paint markers
  • Inside pocket keeps small items secure
  • Has a 30 in. length
Catalog No. 50-006-75982 Supplier MSC Supplier No. 98653470
This welding jacket offers reliable protection for light welding applications.
  • Constructed from 9 oz. flame-resistant treated cotton
  • Welder's collar for enhanced protection against sparks and splatter
  • Equipped with 6 convenient snap buttons, including a collar button
  • Leather reinforcements applied to all snaps
  • Scribe pocket holds welding pencils, soapstone, and paint markers
  • Inside pocket keeps small items secure
  • Has a 30 in. length