Learn More
US Plastic Corporation Black Tamco® Tray 30" L x 24" x 3" Hgt
SDP
Catalog No. NC3815212
Encompass
Available in an array of sizes, these heavy-duty Tamco® Trays are durable, strong, and great for use in a variety of industrial applications. Made of a UV stabilized LLDPEChemical resistantWall thickness: 3/16"Withstands temperature up to 140°FMeets FDA standardsDimensions are nominal
Catalog No. NC3815212 Supplier US Plastic Corporation Supplier No. 17946
Available in an array of sizes, these heavy-duty Tamco® Trays are durable, strong, and great for use in a variety of industrial applications. Made of a UV stabilized LLDPEChemical resistantWall thickness: 3/16"Withstands temperature up to 140°FMeets FDA standardsDimensions are nominal