Learn More
Cayman Chemical Black Tea TheaflavIn 10mg
SDP
Catalog No. 502933542
Encompass_Preferred
A polyphenolic flavonoid with antioxidant and anticancer activities; scavenges singlet oxygen and hydrogen peroxide as well as superoxide and hydroxide radicals in vitro (IC50s = 0.73, 0.49, 14.5, and 38.0 µM, respectively); inhibits the growth of OVCAR-3 and A2780/CP70 human ovarian cancer cell lines, but not the ovarian epithelial cell line IOSE 364 (IC50s = 11.9, 38.5, and >40 µM, respectively)
Catalog No. 50-293-3542 Supplier Cayman Chemical Supplier No. 2512910 MG
A polyphenolic flavonoid with antioxidant and anticancer activities; scavenges singlet oxygen and hydrogen peroxide as well as superoxide and hydroxide radicals in vitro (IC50s = 0.73, 0.49, 14.5, and 38.0 µM, respectively); inhibits the growth of OVCAR-3 and A2780/CP70 human ovarian cancer cell lines, but not the ovarian epithelial cell line IOSE 364 (IC50s = 11.9, 38.5, and >40 µM, respectively)