Learn More
Quality Biological Inc Permanent Black Ink Tube Marker, Fast Drying Black Ink, 5/CS
SDP
Catalog No. 50147442
Encompass_Preferred
Permanent Black Ink Tube Marker, Fast Drying Black Ink, 5/CS
Fast drying, smudge resistant, fine point marker
Ideal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes!
Alcohol/Solvent Resistant: No
Catalog No. 50-147-442 Supplier Quality Biological Inc Supplier No. P886229407
Permanent Black Ink Tube Marker, Fast Drying Black Ink, 5/CS
Fast drying, smudge resistant, fine point marker
Ideal for permanently marking all plastic lab ware, especially centrifuge tubes; even works with frozen tubes!
Alcohol/Solvent Resistant: No