Learn More
Perkin Elmer US LLC Black Tygon R2400 Tubing - 1 ft x 0.25 in O.D., 0.125 in I.D.
Catalog No. 502111994
Encompass_Preferred
Multi-purpose Tygon R2400 tubing is used on a variety of different PerkinElmer instruments. It has an outer diameter 0.25 in, inner diameter 0.125 in, and is black. The tubing is sold by the foot.
Catalog No. 50-211-1994 Supplier Perkin Elmer US LLC Supplier No. 02506515
Multi-purpose Tygon R2400 tubing is used on a variety of different PerkinElmer instruments. It has an outer diameter 0.25 in, inner diameter 0.125 in, and is black. The tubing is sold by the foot.