Learn More
Uline UV Stabilized Nylon Cable Ties, Black
Catalog No. NC2382848
Encompass
Tamperproof ties secure bags, cords and cables permanently. Approved for outdoor use. Made of 6.6 nylon resin. Locks tightly - Won�t stretch or slide. Temperature range -40 to 180 F.
Catalog No. NC2382848 Supplier Uline Supplier No. S12396
Tamperproof ties secure bags, cords and cables permanently. Approved for outdoor use. Made of 6.6 nylon resin. Locks tightly - Won�t stretch or slide. Temperature range -40 to 180 F.