Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuform™ Black/Yellow Marking Tape with Diagonal Stripes

Catalog No. 19169799 Shop All AccuformNMC Products
Change view
Click to view available options
Color:
Black with Yellow Stripes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-169-799 Black with Yellow Stripes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-169-799 Supplier Accuform™ Supplier No. PTM723BKYL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used to mark specific areas and promote safety by identifying potential hazard areas

  • Waterproof and wear-resistant
  • Permanent self-adhesive back with no liner
  • Minor resistance to acids, oils, grease, caustics, alkalis and most solvents
  • Adheres to floors, walls, piping, posts, beams, handrails, stairways, machinery and equipment

Specifications

Width (English) 2 in.
Length (English) 54 ft.
Thickness 8 mil
For Use With (Application) For marking areas and promoting safety
Color Black with Yellow Stripes
Type Marking Tape
Adhesive Type Permanent Adhesive
Material Vinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.