Learn More
MSC Gateway Safety BlackHawk Cap-Mount Earmuff, 24 NRR, Black, 1 ea.
Catalog No. 5000685642
Encompass
The BlackHawk cap-mount earmuff provides robust hearing protection in a dielectric design. Engineered for comfort and utility in demanding work environments, this model integrates directly with hard hats for streamlined use.
  • Features a patented low profile arm for secure cap mounting
  • Equipped with Soft-Seal™ ear cushions for enhanced comfort
  • Provides a noise reduction rating of 24 NRR
  • Tested to ANSI S3.19-1974 standards
  • Manufactured in the USA
Catalog No. 50-006-85642 Supplier MSC Supplier No. 02947596
The BlackHawk cap-mount earmuff provides robust hearing protection in a dielectric design. Engineered for comfort and utility in demanding work environments, this model integrates directly with hard hats for streamlined use.
  • Features a patented low profile arm for secure cap mounting
  • Equipped with Soft-Seal™ ear cushions for enhanced comfort
  • Provides a noise reduction rating of 24 NRR
  • Tested to ANSI S3.19-1974 standards
  • Manufactured in the USA