Learn More
Biocare Medical, LLC BLACKOUT IMPROV RGNT VIAL 7ML
SDP
Catalog No. NC2585268
Encompass
The ONCORE Pro Improv Reagent Vials are user-fillable vials designed to utilize alternate primary antibodies or reagents not offered within the ONCORE reagent line on the ONCORE Automated Slide Staining System.These ONCORE Pro Improv Reagent vials are black-out vials for light-sensitive reagents.
Catalog No. NC2585268 Supplier Biocare Medical, LLC Supplier No. ONCPR104H
The ONCORE Pro Improv Reagent Vials are user-fillable vials designed to utilize alternate primary antibodies or reagents not offered within the ONCORE reagent line on the ONCORE Automated Slide Staining System.These ONCORE Pro Improv Reagent vials are black-out vials for light-sensitive reagents.