Learn More
GA International Blackout Paper Labels 1.5" x 0.5" / 38.1mm x 12.7mm Rectangle White From -54°C to +93°C (-65.2°F to +199.4°F)
Catalog No. 5000571404
Encompass_Preferred
Permanent blackout paper labels with a unique opaque design that allows them to obscure any existing printed information, including serial numbers, barcodes, and text. Perfectly suited for general office use, laboratory, and warehouse applications, these cover-up labels are available in roll format and compatible with all standard thermal-transfer barcode printers.
Catalog No. 50-005-71404 Supplier GA International Supplier No. BOP240C35WH
Permanent blackout paper labels with a unique opaque design that allows them to obscure any existing printed information, including serial numbers, barcodes, and text. Perfectly suited for general office use, laboratory, and warehouse applications, these cover-up labels are available in roll format and compatible with all standard thermal-transfer barcode printers.