Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carson™ BlackWave Waterproof Monocular
SDPSCI_ED

Catalog No. S13545 Shop All Carson Optical Products
Change view
Click to view available options
Field of View:
273 ft at 1,000 yards
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Field of View
S13545 273 ft at 1,000 yards
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13545 Supplier Carson™ Supplier No. WM025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carson™ BlackWave Waterproof Monocular is a 10 x 25 mm waterproof monocular with a rugged design.

  • Ideal for outdoor adventures like camping, hiking, concerts and sporting events as it will stand up to the harshest of elements.
  • Lightweight and portable
  • Close focus feature of just 4 ft. to meet multi-functional viewing needs
  • No Fault, No Hassle Warranty.

Specifications

Magnification 10x
Lens Coating BK-7
Includes Pouch, neck strap and lens cloth
Field of View 273 ft at 1,000 yards
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.