Learn More
Discovery Life Sciences Bladder Ca Act Tx IO PBMC
SDP
Catalog No. 5000804088
Encompass
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays
Catalog No. 50-008-04088 Supplier Discovery Life Sciences Supplier No. 121010
PBMCs Peripheral Blood Mononuclear Cells are immune cells isolated from whole blood and viably cryopreserved providing a foundational material for immunology studies cell therapy research and functional immune assays