Learn More
Discovery Life Sciences Bladder Cancer PD L1 DTC
SDP
Catalog No. 5000803977
Encompass
DTC Dissociated Tumor and Tissue Cells start as fresh solid tumor tissue collected during surgery are enzymatically and mechanically dissociated into single cells and are delivered as viably cryopreserved cells for functional assays cell culture and single cell genomics
Catalog No. 50-008-03977 Supplier Discovery Life Sciences Supplier No. 321110.3
DTC Dissociated Tumor and Tissue Cells start as fresh solid tumor tissue collected during surgery are enzymatically and mechanically dissociated into single cells and are delivered as viably cryopreserved cells for functional assays cell culture and single cell genomics