Learn More
ATCC Bladder Epithelial Cell Growth Kit
Catalog No. 502382219
Encompass
The kit contains components that when added to Bladder Epithelial Cell Basal Medium (ATCC PCS-420-032) create a complete ATCC Primary Cell Solutions™ culture environment for epithelial cells derived from normal human bladder tissue. The serum-free medium formulation is designed to support normal bladder epithelial cell morphology as well as promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins or other substrates are required. The final concentration for each component in complete bladder epithelial cell growth medium is as follows: L-Glutamine: 6 mM Extract P: 0.4% Epinephrine: 1.0 μM rh EGF: 5 ng/mL Hydrocortisone: 100 ng/mL Insulin: 5 μg/mL Apo-transferrin: 5 μg/mL rh TGF-α: 0.5 ng/mL rh KGF: 5 ng/mL
Catalog No. 50-238-2219 Supplier ATCC Supplier No. PCS420042
Add to Cart
Add to Cart
The kit contains components that when added to Bladder Epithelial Cell Basal Medium (ATCC PCS-420-032) create a complete ATCC Primary Cell Solutions™ culture environment for epithelial cells derived from normal human bladder tissue. The serum-free medium formulation is designed to support normal bladder epithelial cell morphology as well as promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins or other substrates are required. The final concentration for each component in complete bladder epithelial cell growth medium is as follows: L-Glutamine: 6 mM Extract P: 0.4% Epinephrine: 1.0 μM rh EGF: 5 ng/mL Hydrocortisone: 100 ng/mL Insulin: 5 μg/mL Apo-transferrin: 5 μg/mL rh TGF-α: 0.5 ng/mL rh KGF: 5 ng/mL