Learn More
Med Vet International Surgical Blade Size 22 Sterile Bard Parker 50/Box
Catalog No. 502831446
Encompass_Preferred
(BD Medical) The superior carbon steel construction of these sterile surgical blades delivers consistent sharpness. Carbon blades feature the Rib-Back design, which enhances feel and improves cutting control. These blades have consistent sharpness, control and strength surgeons demand and are uniquely packaged for safety and ease of use.
Catalog No. 50-283-1446 Supplier Med Vet International Supplier No. 371122
(BD Medical) The superior carbon steel construction of these sterile surgical blades delivers consistent sharpness. Carbon blades feature the Rib-Back design, which enhances feel and improves cutting control. These blades have consistent sharpness, control and strength surgeons demand and are uniquely packaged for safety and ease of use.