Learn More
Med Vet International Pivoting Clipper Blade Assembly 3M Single Use, Nonsterile, 9660, 50/Case
Catalog No. 502831419
Encompass_Preferred
For use with 3M™ Surgical Clipper 9661 Conforms to contours of patient's body Adjustable head allows clipping with a pull or push motion Reduces risk of cross-contamination with single-use blades Blade easily snaps-on and pops-off
Catalog No. 50-283-1419 Supplier Med Vet International Supplier No. 347468CS
For use with 3M™ Surgical Clipper 9661 Conforms to contours of patient's body Adjustable head allows clipping with a pull or push motion Reduces risk of cross-contamination with single-use blades Blade easily snaps-on and pops-off