Learn More
Electron Microscopy Sciences Razor Blade Double Edge Coated Blade Washed
SDP
Supplier:  Electron Microscopy Sciences 72001WA
Encompass_Preferred
Stainless Steel "PTFE" coated. PERSONNA brand .004" thick, wrapped individually. Washed and degreased reducing foreign material. Clean room classified.
Catalog No. 50-192-7063