Learn More
StatLab Blade Remover
SDP
Catalog No. 22035379
Change view
Click to view available options
Size:
# 22
Catalog No. Size
22-035-379 # 22
1 options
Catalog No. 22-035-379 Supplier StatLab Supplier No. SL92500
Add to Cart
Add to Cart
Convenient method for removal of no. 22 scalpel blades
  • Insert blade and handle into the slot, press and pull
  • Blade falls safely into a compartment that stores up to 100 blades
  • Sold individually
Specifications
Product Type Scalpel Blade Remover
Size # 22