Learn More
Mopec Blade Remover, Disposable, 10 pack 
SDP
Catalog No. 22444269 Shop All Mopec Products
Change view
Click to view available options
Disposable:
Single-use
Catalog No. Disposable
22-444-269 Single-use
1 options
Catalog No. 22-444-269 Supplier Mopec Supplier No. AJ116
Add to Cart
Add to Cart
Mopec Blade remover designed to safely remove dull contaminated blade from scalpel handle.
  • Easy to use
  • Autoclavable
  • Holds 100 blades each
  • Not compatible with double sided blades
Specifications
Product Type Blade Remover
For Use With (Application) Scalpel Blade
Disposable Single-use
Autoclavable Autoclavable
Quantity 10/Pk.