Learn More
Med Vet International Scalpel Blades #10 Size, 100/Box
Catalog No. 502830457
Encompass_Preferred
Premiere #10 Blades are made of high carbon steel, individually foil wrapped and sterilized by gamma radiation. 100 per box.
Catalog No. 50-283-0457 Supplier Med Vet International Supplier No. 9310
Premiere #10 Blades are made of high carbon steel, individually foil wrapped and sterilized by gamma radiation. 100 per box.