Learn More
Med Vet International Miltex Stainless Steel Sterile Surgical Blades, #12, 100/box, Individually Wrapped
Catalog No. 503045639
Encompass_Preferred
The Miltex 4-312 #12 stainless steel sterile surgical blades. Features: Razor-sharp cutting edge for superior cutting and clean incisions Sterile, 5-year shelf life Individually packaged in foil peel packs Convenience of disposables, single-use Sizes clearly marked on each blade Each box contains 100 blades.
Catalog No. 50-304-5639 Supplier Med Vet International Supplier No. MIL4312
The Miltex 4-312 #12 stainless steel sterile surgical blades. Features: Razor-sharp cutting edge for superior cutting and clean incisions Sterile, 5-year shelf life Individually packaged in foil peel packs Convenience of disposables, single-use Sizes clearly marked on each blade Each box contains 100 blades.