Learn More
Med Vet International Miltex Carbon Steel Sterile Surgical Blades, #15, 100/Box, Individually Wrapped
Catalog No. 502829715
Encompass_Preferred
The Miltex 4-115 #15 carbon steel sterile surgical blades. Features: Razor-sharp cutting edge for superior cutting and clean incisions Sterile, 5-year shelf life Individually packaged in foil peel packs Convenience of disposables, single-use Sizes clearly marked on each blade Each box contains 100 blades.
Catalog No. 50-282-9715 Supplier Med Vet International Supplier No. MIL4115
The Miltex 4-115 #15 carbon steel sterile surgical blades. Features: Razor-sharp cutting edge for superior cutting and clean incisions Sterile, 5-year shelf life Individually packaged in foil peel packs Convenience of disposables, single-use Sizes clearly marked on each blade Each box contains 100 blades.