Learn More
Med Vet International Surgical Clipper Blade 3M, Nonsterile, 9680, 50/Case
Catalog No. 502829831
Encompass_Preferred
The 3M™ surgical clipper blade facilitates effective and comfortable patient hair removal A 36.4-millimeter blade width provides maximum coverage for high efficiency Its universal design accommodates a variety of hair clipping needs for all areas of the body A one-blade approach saves on additional costs Its cutting position optimizes safety to protect the skin and prevent potential infections The slide on and off mechanism makes the blade easy to assemble with one hand
Catalog No. 50-282-9831 Supplier Med Vet International Supplier No. 836200CS
The 3M™ surgical clipper blade facilitates effective and comfortable patient hair removal A 36.4-millimeter blade width provides maximum coverage for high efficiency Its universal design accommodates a variety of hair clipping needs for all areas of the body A one-blade approach saves on additional costs Its cutting position optimizes safety to protect the skin and prevent potential infections The slide on and off mechanism makes the blade easy to assemble with one hand