Learn More
Med Vet International Size 10 Carbon Steel Sterile Surgical Blades, 100/Box
Supplier:  Med Vet International ZB10
Encompass_Preferred
The #10 blade with its curved cutting edge is one of the more traditional blade shapes and is used generally for making varying sizes of incision in skin and muscle. The #10 is often utilized in more specialized surgeries such as for harvesting the radial artery during a coronary artery bypass operation, opening the bronchus during thoracic surgery, uterine incisions for a C-Section and for Inguinal hernia repair. The #10 fits handles 3, 3L, 3 Graduated, 5B, 7, 9 and B3. Years of research and engineering have gone into the production of these high-quality, world-standard surgical blades ... the definite choice of leading surgeons and hospitals worldwide. These blades offer excellent value and significant savings.
Catalog No. 50-283-2169