Learn More
Med Vet International #22 Carbon Steel Surgical Blades, 100/Box, 1, 000/Case
Supplier:  Med Vet International #22CS
Encompass_Preferred
The #22 is a large version of the #10 blade with a curved cutting edge and a flat, unsharpened back edge. Used for skin incisions in both cardiac and thoracic surgery and to cut the bronchus in lung resection surgery. The #22 blade fits handles 4, 4L, 4 Graduated and 6B.Years of research and engineering have gone into the production of these high quality world standard surgical blades; the definite choice of leading surgeons and hospitals worldwide. This blades offer excellent value and significant savings.
Catalog No. 50-118-0248