Learn More
Med Vet International #15 Carbon Steel Surgical Blade, 100/Box
Catalog No. 502522588
Encompass_Preferred
The #15 blade has a small curved cutting edge and is the most popular blade shape ideal for making short and precise incisions. It is utilized in a variety of surgical procedures including the excision of a skin lesion or recurrent sebaceous cyst and for opening coronary arteries. Years of research and engineering have gone into the production of these high-quality, world-standard surgical blades — the definite choice of leading surgeons and hospitals worldwide. Our #15 blades offer excellent value and significant savings.
Catalog No. 50-252-2588 Supplier Med Vet International Supplier No. 15
The #15 blade has a small curved cutting edge and is the most popular blade shape ideal for making short and precise incisions. It is utilized in a variety of surgical procedures including the excision of a skin lesion or recurrent sebaceous cyst and for opening coronary arteries. Years of research and engineering have gone into the production of these high-quality, world-standard surgical blades — the definite choice of leading surgeons and hospitals worldwide. Our #15 blades offer excellent value and significant savings.