Learn More
Med Vet International Miltex Blade-Safe Surgical Blade Remover
Catalog No. 502831848
Encompass_Preferred
The Miltex 4-100 Blade-Safe surgical blade remove is made of OR grade German stainless steel. It is designed to safely remove blades from handles while protecting hands. Works with any style of handle.
Catalog No. 50-283-1848 Supplier Med Vet International Supplier No. MIL4100
The Miltex 4-100 Blade-Safe surgical blade remove is made of OR grade German stainless steel. It is designed to safely remove blades from handles while protecting hands. Works with any style of handle.