Learn More
Electron Microscopy Sciences Lance Stainless Steel Blade Size 15, Sterile
SDP
Catalog No. 501928430
Encompass_Preferred
These disposable scalpels are sterile, sealed indiviually in trays and boxed in units of 10. The stainless steal blades are mounted on Polystyrene handles.
Material: Stainless Steel
Sterile: Sterile
This item is not returnable. View return policy
Catalog No. 50-192-8430 Supplier Electron Microscopy Sciences Supplier No. 7205409
These disposable scalpels are sterile, sealed indiviually in trays and boxed in units of 10. The stainless steal blades are mounted on Polystyrene handles.
Material: Stainless Steel
Sterile: Sterile