Learn More
Electron Microscopy Sciences Lance Carbon Steel Blade Size 23, Sterile
SDP
Catalog No. 501928435
Encompass_Preferred
These disposable scalpels are sterile, sealed indiviually in trays and boxed in units of 10. The stainless steal blades are mounted on Polystyrene handles.
Material: Carbon Steel
Sterile: Sterile
This item is not returnable. View return policy
Catalog No. 50-192-8435 Supplier Electron Microscopy Sciences Supplier No. 7205414
These disposable scalpels are sterile, sealed indiviually in trays and boxed in units of 10. The stainless steal blades are mounted on Polystyrene handles.
Material: Carbon Steel
Sterile: Sterile