Learn More
Ted Pella Inc PELCO Blades for use with Brain Matrices, pkg/25
SDP
Catalog No. NC9016444
Encompass
This single edge thin, extra-long razor blade is made from uncoated carbon steel. The blade is 120mm long, 20mm wide and 0.12mm thick (4.72 x 0.78 x 0.005"). It is double beveled for increased strength and durability. The blades are lightly protected with mineral oil for corrosion protection. It is recommended to clean each blade with Isopropanol (Prod. No. 18242) before use. This long sharp thin blade is developed specifically for use with large brain matrices.
Catalog No. NC9016444 Supplier Ted Pella Inc Supplier No. 12196
This single edge thin, extra-long razor blade is made from uncoated carbon steel. The blade is 120mm long, 20mm wide and 0.12mm thick (4.72 x 0.78 x 0.005"). It is double beveled for increased strength and durability. The blades are lightly protected with mineral oil for corrosion protection. It is recommended to clean each blade with Isopropanol (Prod. No. 18242) before use. This long sharp thin blade is developed specifically for use with large brain matrices.