Learn More
Electron Microscopy Sciences Opticut™ LP Low Profile Blades, PTFE Coated, .012" thick
SDP
Catalog No. 501928596
Encompass_Preferred
Ultra-durable and long-lasting, these blades are ideal for both paraffin and frozen sections.
Type: PTFE
Thickness: .012"
Catalog No. 50-192-8596 Supplier Electron Microscopy Sciences Supplier No. 6305702
Ultra-durable and long-lasting, these blades are ideal for both paraffin and frozen sections.
Type: PTFE
Thickness: .012"