Learn More
Med Vet International Dynarex Medicut Blades, #23, 100/Box
Catalog No. 502830535
Encompass_Preferred
Dynarex Medi-Cut Blades are crafted from premium-quality stainless steel for optimal performance. The Blades feature a finely honed cutting edge to allow for precise and effective surgical incisions. The Medi-Cut Blades are sterile and individually foil packaged for optimal hygiene. Dynarex Medi-Cut Blades are high-quality stainless-steel blades designed with a finely honed cutting edge for improved performance and precision. 
Catalog No. 50-283-0535 Supplier Med Vet International Supplier No. 4143
Dynarex Medi-Cut Blades are crafted from premium-quality stainless steel for optimal performance. The Blades feature a finely honed cutting edge to allow for precise and effective surgical incisions. The Medi-Cut Blades are sterile and individually foil packaged for optimal hygiene. Dynarex Medi-Cut Blades are high-quality stainless-steel blades designed with a finely honed cutting edge for improved performance and precision. 