Learn More
DR Instruments Blades for Scalpel Handles
Description
- Disposable Surgical blades designed for scalpel handles #3 and #4, offering effortless installation and removal.
- These exceptionally sharp surgical blades facilitate the precise cutting of preserved specimens.
- Crafted from high-quality carbon steel, they are compatible with scalpel handles No. 3 and 4.
Specifications
Specifications
| Blade Material | Carbon Steel |
| Product Type | Blade |
| Size | # 12 |
| For Use With (Equipment) | Scalpel Handle # 3 |
| Disposable | Single-use |