Learn More
Med Vet International Miltex Sterile Carbon Steel Surgical Blades #12, 100/Box, 4-112
Supplier:  Med Vet International MIL4112
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Sterile Carbon Steel Surgical Blades. Surgical blades can be used for surgery, anatomical dissection, podiatry and various arts and crafts. Specifications: #12, 100/Box
Catalog No. 50-283-1572