Learn More
Electron Microscopy Sciences Swann-Morton Carbon Steel Blades Size 11, Non-Sterile
SDP
Supplier:  Electron Microscopy Sciences 7205317
Encompass_Preferred
Swann-Morton blades feature 200 percent visual inspection and high surface finish. They are packaged with VCI paper to protect the blade from corrosion and have a cellophane overwrap. The narrow grinding angle results in increased initial keenness. Wrapped 5 to a package.
Material: Carbon Steel
Sterile: Non-Sterile
Catalog No. 50-192-8352