Learn More
Electron Microscopy Sciences Swann-Morton Stainless Steel Blades Size 26, Sterile
SDP
Supplier:  Electron Microscopy Sciences 7205381
Encompass_Preferred
Swann-Morton blades feature 200 percent visual inspection and high surface finish. They are packaged with VCI paper to protect the blade from corrosion and have a cellophane overwrap. The narrow grinding angle results in increased initial keenness. Individually wrapped.
Material: Stainless Steel
Sterile: Sterile
Catalog No. 50-192-8415