Learn More
Electron Microscopy Sciences Swann-Morton Stainless Steel Blades Size E11, Sterile
SDP
Catalog No. 501928358
Encompass_Preferred
Swann-Morton blades feature 200 percent visual inspection and high surface finish. They are packaged with VCI paper to protect the blade from corrosion and have a cellophane overwrap. The narrow grinding angle results in increased initial keenness. Individually wrapped.
Material: Stainless Steel
Sterile: Sterile
Catalog No. 50-192-8358 Supplier Electron Microscopy Sciences Supplier No. 7205323
Swann-Morton blades feature 200 percent visual inspection and high surface finish. They are packaged with VCI paper to protect the blade from corrosion and have a cellophane overwrap. The narrow grinding angle results in increased initial keenness. Individually wrapped.
Material: Stainless Steel
Sterile: Sterile