Learn More
StatLab Bladex Blade Exchange Device
SDP
Catalog No. 22000013
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
22-000-013 1 Ea.
1 options
Catalog No. 22-000-013 Supplier StatLab Supplier No. BLDX2260DS
Bladex Blade Exchange Device
  • A one-of-a-kind product which removes used blades
  • Collects blades in a SHARPS container below the device
  • Attaches a fresh clean blade to scalpels
  • Improve safety and convenience in your lab
Specifications
Type Accessioning and Sample Transfer Instrument
Quantity 1 Ea.