Learn More
StatLab Bladex Reusable Blade Handle
SDP
Catalog No. 22000039
Change view
Click to view available options
Size:
No. 22 and No. 60
Catalog No. Size
22-000-039 No. 22 and No. 60
1 options
Catalog No. 22-000-039 Supplier StatLab Supplier No. HNDL2260
A safe and convenient method for exchanging a used scalpel blade with a fresh blade
  • Compatible with Bladex Safety Blade Exchange Device
  • The entire process takes 2-3 seconds
  • During this one-handed operation, the user does not have to touch a blade and the used blade is disposed of in a sealed biohazard sharps container
Specifications
Product Type Scalpel Handle
Size No. 22 and No. 60
Disposable Reusable
Sterility Non-sterile
Autoclavable Not Autoclavable
Graduated No