Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BLaER1 Human B-cell Precursor Leukemia Cell Line

Catalog No. SCC177 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
≥ 1 x 106 cells/vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCC177 ≥ 1 x 106 cells/vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCC177 Supplier MilliporeSigma™ Supplier No. SCC177
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BLaER1 human B-cell precursor leukemia cell line may be transdifferentiated and used to model human monocytes.

  • Induction of differentiation is a therapeutic approach for cancer
  • Treatment of leukemias with differentiation-inducing factors in combination with pro-apoptotic compounds has been shown to be highly effective
  • Ectopic expression of the transcription factor C/EBPα causes reprogramming of leukemia cells, resulting in expression of a normal cell phenotype
  • The BLaER1 cell line is a tamoxifen-inducible derivative of the RCH-ACV B-cell leukemia cell line genetically modified using a retrovirus vector to express C/EBPα fused with the estrogen receptor hormone binding domain (ER) and GFP
  • A study demonstrated that BLaER1 cells could efficiently convert into cells exhibiting increased adherent, phagocytic and quiescent properties with a transcriptome resembling normal macrophages
  • Cells retained their phenotype even when C/EBPα was inactivated, a characteristic of genuine cell reprogramming
  • C/EBPα induction also impaired cell tumorigenicity after transplantation into immunodeficient mice
  • BLaER1 is thus a model for highly efficient transdifferentiation of B-cell-derived human cancer cells
  • The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality
  • Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services
  • Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, Chinese hamster, Golden Syrian hamster, and non-human primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
  • Cells are negative for mycoplasma contamination
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in liquid nitrogen
Format Vial
Quantity ≥ 1 x 106 cells/vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.