Learn More
VEOLIA WTS TOC KHP Set, 1 mg/l
Catalog No. NC0223222
Encompass
TOC check standards set for Sievers 900 Series. Includes: (1) Blank water and (2) 1 mg C/L TOC standard (as KHP)
Catalog No. NC0223222 Supplier VEOLIA WTS Supplier No. STD 9004601
TOC check standards set for Sievers 900 Series. Includes: (1) Blank water and (2) 1 mg C/L TOC standard (as KHP)