Learn More
OHAUS Blank Foam Inserts 2
Catalog No. 01804424 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-424 Vortex Attachment
1 options
Catalog No. 01-804-424 Supplier OHAUS Supplier No. 83041432
Add to Cart
Add to Cart
Two high-density Foam Inserts are customizable by the user for odd-shaped vessels.
  • The insert fits into the Recessed Platform 504-0039-00, for use with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model Vortex-Genie 1, Vortex-Genie Pulse and the Vortex-Genie 2